Läs mer
enQuireBio™ Recombinant Human Butyrophilin subfamily 2 member A2 Protein
Produktkod. p-7154634
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16021811
Brand: enQuireBio™ QP7953ec10ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Homo sapiens (Human) Butyrophilin subfamily 2 member A2, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_001184166.1
Gen-ID (Entrez) 10385
Namn Butyrophilin subfamily 2 member A2 Protein
Kvantitet 10 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol BTN2A2
Protein Längd Partial (see sequence information for more details). of Isoform 2
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag His-SUMO
Sekvens QFTVVGPANP ILAMVGENTT LRCHLSPEKN AEDMEVRWFR SQFSPAVFVY KGGRERTEEQ MEEYRGRITF VSKDINRGSV ALVIHNVTAQ ENGIYRCYFQ EGRSYDEAIL RLVVAGLGSK PLIEIKAQED GSIWLECISG GWYPEPLTVW RDPYGEVVPA LKEVSIADAD GLFMVTTAVI IRDKYVRNVS CSVNNTLLGQ EKETVIFIPE SFMPSASPWM VALAVILTAS
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.