missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-Myc-binding Protein
A DNA sequence encoding the Homo sapiens (Human) C-Myc-binding protein, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_036465.2 |
---|---|
Gene ID (Entrez) | 26292 |
Name | C-Myc-binding Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16042171
|
enQuireBio™
QP8016-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16062171
|
enQuireBio™
QP8016-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16052171
|
enQuireBio™
QP8016-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072171
|
enQuireBio™
QP8016-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082171
|
enQuireBio™
QP8016-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092171
|
enQuireBio™
QP8016-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_036465.2 | |
C-Myc-binding Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
26292 | |
Research Use Only | |
MYCBP | |
Recombinant Protein | |
E. coli | |
AHYKAADSKR EQFRRYLEKS GVLDTLTKVL VALYEEPEKP NSALDFLKHH LGAATPENPE IELLRLELAE MKEKYEAIVE ENKKLKAKLA QYEPPQEEKR AE | |
Greater than 90% as determined by SDS-PAGE. |