missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-type lectin domain family 4 member C Protein
A DNA sequence encoding the Homo sapiens (Human) C-type lectin domain family 4 member C, was expressed in the hosts and tags indicated.
4825.00 SEK - 13070.00 SEK
Specifications
Accession Number | NP_569708.1 |
---|---|
Gene ID (Entrez) | 170482 |
Name | C-type lectin domain family 4 member C Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16072031
|
enQuireBio™
QP7991-MA-20UG |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082031
|
enQuireBio™
QP7991-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062031
|
enQuireBio™
QP7991-MA-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_569708.1 | |
C-type lectin domain family 4 member C Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
170482 | |
Research Use Only | |
CLEC4C | |
Recombinant Protein | |
Mammal | |
NFMYSKTVKR LSKLREYQQY HPSLTCVMEG KDIEDWSCCP TPWTSFQSSC YFISTGMQSW TKSQKNCSVM GADLVVINTR EEQDFIIQNL KRNSSYFLGL SDPGGRRHWQ WVDQTPYNEN VTFWHSGEPN NLDERCAIIN FRSSEEWGWN DIHCHVPQKS ICKMKKIYI | |
Greater than 80% as determined by SDS-PAGE. |