missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-X-C chemokine receptor type 4 Protein
A DNA sequence encoding the Homo sapiens (Human) C-X-C chemokine receptor type 4, was expressed in the hosts and tags indicated.
5820.00 SEK - 19155.00 SEK
Specifications
Accession Number | NP_001008540.1 |
---|---|
Gene ID (Entrez) | 7852 |
Name | C-X-C chemokine receptor type 4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059071
|
enQuireBio™
QP9950-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069071
|
enQuireBio™
QP9950-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049071
|
enQuireBio™
QP9950-IV-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001008540.1 | |
C-X-C chemokine receptor type 4 Protein | |
Not determined. | |
Full Length Transmembrane Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7852 | |
Research Use Only | |
CXCR4 | |
Recombinant Protein | |
E. coli | |
MEGISIYTSD NYTEEMGSGD YDSMKEPCFR EENANFNKIF LPTIYSIIFL TGIVGNGLVI LVMGYQKKLR SMTDKYRLHL SVADLLFVIT LPFWAVDAVA NWYFGNFLCK AVHVIYTVNL YSSVLILAFI SLDRYLAIVH ATNSQRPRKL LAEKVVYVGV WIPALLLTIP DFIFANVSEA DDRYICDRFY PNDLWVVVFQ FQHIMVGLIL PGIVILSCYC IIISKLSHSK GHQKRKALKT TVILILAFFA CWLPYYIGIS IDSFILLEII KQGCEFENTV HKWISITEAL AFFHCCLNPI LYAFLGAKFK TSAQHALTSV SRGSSLKILS KGKRGGHSSV STESESSSFH SS | |
Greater than 80% as determined by SDS-PAGE. |