missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-X-C chemokine receptor type 4 Protein
A DNA sequence encoding the Homo sapiens (Human) C-X-C chemokine receptor type 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9950-iv-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001008540.1 | |
C-X-C chemokine receptor type 4 Protein | |
Research Use Only | |
CXCR4 | |
Recombinant Protein | |
E. coli | |
MEGISIYTSD NYTEEMGSGD YDSMKEPCFR EENANFNKIF LPTIYSIIFL TGIVGNGLVI LVMGYQKKLR SMTDKYRLHL SVADLLFVIT LPFWAVDAVA NWYFGNFLCK AVHVIYTVNL YSSVLILAFI SLDRYLAIVH ATNSQRPRKL LAEKVVYVGV WIPALLLTIP DFIFANVSEA DDRYICDRFY PNDLWVVVFQ FQHIMVGLIL PGIVILSCYC IIISKLSHSK GHQKRKALKT TVILILAFFA CWLPYYIGIS IDSFILLEII KQGCEFENTV HKWISITEAL AFFHCCLNPI LYAFLGAKFK TSAQHALTSV SRGSSLKILS KGKRGGHSSV STESESSSFH SS | |
Greater than 80% as determined by SDS-PAGE. |
7852 | |
10 μg | |
Not determined. | |
Full Length Transmembrane Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |