missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cadherin-12 Protein
A DNA sequence encoding the Homo sapiens (Human) Cadherin-12, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8756-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001304156.1 | |
Cadherin-12 Protein | |
Research Use Only | |
CDH12 | |
Recombinant Protein | |
E. coli | |
WVWNQFFVLE EYVGSEPQYV GKLHSDLDKG EGTVKYTLSG DGAGTVFTID ETTGDIHAIR SLDREEKPFY TLRAQAVDIE TRKPLEPESE FIIKVQDIND NEPKFLDGPY VATVPEMSPV GAYVLQVKAT DADDPTYGNS ARVVYSILQG QPYFSIDPKT GVIRTALPNM DREVKEQYQV LIQAKDMGGQ LGGLAGTTIV NITLTDVNDN PPRFPKSIFH LKVPESSPIG SAIGRIRAVD PDFGQNAEIE YNIVPGDGGN LFDIVTDEDT QEGVIKLKKP LDFETKKAYT FKVEASNLHL DHRFHSAGPF KDTATVKISV LDVDEPPVFS KPLYTMEVYE DTPVGTIIGA VTAQDLDVGS SAVRYFIDWK SDGDSYFTID GNEGTIATNE LLDRESTAQY NFSIIASKVS NPLLTSKVNI LINVLDVNEF PPEISVPYET AVCENAKPGQ IIQIVSAADR DLSPAGQQFS FRLSPEAAIK PNFTVRDFRN NTAGIETRRN GYSRRQQELY FLPVVIEDSS YPVQSSTNTM TIRVCRCDSD GTILSCNVEA | |
Greater than 90% as determined by SDS-PAGE. |
1010 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |