Läs mer
enQuireBio™ Recombinant Human Calcium-dependent phospholipase A2 Protein
Produktkod. 16028111
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16028111
Brand: enQuireBio™ QP9333ye200ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Homo sapiens (Human) Calcium-dependent phospholipase A2, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_000920.1
Gen-ID (Entrez) 5322
Namn Calcium-dependent phospholipase A2 Protein
Kvantitet 200 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol PLA2G5
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art Yeast
Protein Tag His
Sekvens GLLDLKSMIE KVTGKNALTN YGFYGCYCGW GGRGTPKDGT DWCCWAHDHC YGRLEEKGCN IRTQSYKYRF AWGVVTCEPG PFCHVNLCAC DRKLVYCLKR NLRSYNPQYQ YFPNILCS
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.