missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Carbonyl reductase family member 4 Protein

A DNA sequence encoding the Homo sapiens (Human) Carbonyl reductase family member 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7969-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061901

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_116172.2
Carbonyl reductase family member 4 Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
84869
Greater than 80% as determined by SDS-PAGE.
Research Use Only
CBR4
Recombinant Protein
E. coli
MDKVCAVFGG SRGIGRAVAQ LMARKGYRLA VIARNLEGAK AAAGDLGGDH LAFSCDVAKE HDVQNTFEEL EKHLGRVNFL VNAAGINRDG LLVRTKTEDM VSQLHTNLLG SMLTCKAAMR TMIQQQGGSI VNVGSIVGLK GNSGQSVYSA SKGGLVGFSR ALAKEVARKK IRVNVVAPGF VHTDMTKDLK EEHLKKNIPL GRFGETIEVA HAVVFLLESP YITGHVLVVD GGLQLIL
Videos
SDS
Documents