missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cardiac phospholamban Protein

A DNA sequence encoding the Homo sapiens (Human) Cardiac phospholamban, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9335-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058121

  • 19700.00 SEK / 500µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002658.1
Cardiac phospholamban Protein
Research Use Only
P26678
Recombinant Protein
Yeast
MEKVQYLTRS AIRRASTIEM PQQARQKLQN LFINFCLILI CLLLICIIVM LL
Greater than 90% as determined by SDS-PAGE.
5350
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers