missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Casein kinase I isoform alpha-like Protein
A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform alpha-like, was expressed in the hosts and tags indicated.
2430.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_660204.2 |
---|---|
Gene ID (Entrez) | 122011 |
Name | Casein kinase I isoform alpha-like Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051501
|
enQuireBio™
QP7904-EC-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071501
|
enQuireBio™
QP7904-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061501
|
enQuireBio™
QP7904-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081501
|
enQuireBio™
QP7904-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091501
|
enQuireBio™
QP7904-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001511
|
enQuireBio™
QP7904-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_660204.2 | |
Casein kinase I isoform alpha-like Protein | |
Not determined. | |
Full Length of BC028723 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
122011 | |
Research Use Only | |
CSNK1A1L | |
Recombinant Protein | |
E. coli | |
MTNNSGSKAE LVVGGKYKLV RKIGSGSFGD VYLGITTTNG EEVAVKLESQ KVKHPQLLYE SKLYTILQGG VGIPHMHWYG QEKDNNVLVM DLLGPSLEDL FNFCSRRFTM KTVLMLADQM ISRIEYVHTK NFLHRDIKPD NFLMGTGRHC NKLFLIDFGL AKKYRDNRTR QHIPYREDKH LIGTVRYASI NAHLGIEQSR RDDMESLGYV FMYFNRTSLP WQGLKAMTKK QKYEKISEKK MSTPVEVLCK GFPAEFAMYL NYCRGLRFEE VPDYMYLRQL FRILFRTLNH QYDYTFDWTM LKQKAAQQAA SSSGQGQQAQ TQTGKQTEKN KNNVKDN | |
Greater than 80% as determined by SDS-PAGE. |