missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Caspase-8 Protein
A DNA sequence encoding the Homo sapiens (Human) Caspase-8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8921-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
Caspase-8 Protein | |
Research Use Only | |
CASP8 | |
Recombinant Protein | |
E. coli | |
SESQTLDKVY QMKSKPRGYC LIINNHNFAK AREKVPKLHS IRDRNGTHLD AGALTTTFEE LHFEIKPHDD CTVEQIYEIL KIYQLMDHSN MDCFICCILS HGDKGIIYGT DGQEAPIYEL TSQFTGLKCP SLAGKPKVFF IQACQGDNYQ KGIPVETDSE EQPYLEMDLS SPQTRYIPDE ADFLLGMATV NNCVSYRNPA EGTWYIQSLC QSLRERCPRG DDILTILTEV NYEVSNKDDK KNMGKQMPQP TFTLRKKLVF PSD | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |