missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL20 / MIP-3 alpha Protein

A DNA sequence encoding the Homo sapiens (Human) CCL20 / MIP-3 alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8520-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065081

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001123518.1
CCL20 / MIP-3 alpha Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6364
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CCL20
Recombinant Protein
E. coli
ASNFDCCLGY TDRILHPKFI VGFTRQLANE GCDINAIIFH TKKKLSVCAN PKQTWVKYIV RLLSKKVKN
Videos
SDS
Documents