missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCR4-NOT transcription complex subunit 8 Protein
A DNA sequence encoding the Homo sapiens (Human) CCR4-NOT transcription complex subunit 8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8105-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001288002.1 | |
CCR4-NOT transcription complex subunit 8 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9337 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
CNOT8 | |
Recombinant Protein | |
E. coli | |
MPAALVENSQ VICEVWASNL EEEMRKIREI VLSYSYIAMD TEFPGVVVRP IGEFRSSIDY QYQLLRCNVD LLKIIQLGLT FTNEKGEYPS GINTWQFNFK FNLTEDMYSQ DSIDLLANSG LQFQKHEEEG IDTLHFAELL MTSGVVLCDN VKWLSFHSGY DFGYMVKLLT DSRLPEEEHE FFHILNLFFP SIYDVKYLMK SCKNLKGGLQ EVADQLDLQR IGRQHQAGSD SLLTGMAFFR MKELFFEDSI DDAKYCGRLY GLGTGVAQKQ NEDVDSAQEK MSILAIINNM QQ |