missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD147 / EMMPRIN / Basigin Protein
A DNA sequence encoding the Homo sapiens (Human) CD147 / EMMPRIN / Basigin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8729-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001309172.1 | |
CD147 / EMMPRIN / Basigin Protein | |
Research Use Only | |
BSG | |
Recombinant Protein | |
E. coli | |
EPGTVFTTVE DLGSKILLTC SLNDSATEVT GHRWLKGGVV LKEDALPGQK TEFKVDSDDQ WGEYSCVFLP EPMGTANIQL HGPPRVKAVK SSEHINEGET AMLVCKSESV PPVTDWAWYK ITDSEDKALM NGSESRFFVS SSQGRSELHI ENLNMEADPG QYRCNGTSSK GSDQAIITLR VRSH | |
Greater than 90% as determined by SDS-PAGE. |
682 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |