missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD171 / N-CAML1 / L1CAM Protein
A DNA sequence encoding the Homo sapiens (Human) CD171 / N-CAML1 / L1CAM, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8574-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000416.1 | |
CD171 / N-CAML1 / L1CAM Protein | |
Research Use Only | |
L1CAM | |
Recombinant Protein | |
E. coli | |
EAIVREGGTM ALSGISDFGN ISATAGENYS VVSWVPKEGQ CNFRFHILFK ALGEEKGGAS LSPQYVSYNQ SSYTQWDLQP DTDYEIHLFK ERMFRHQMAV KTNGTGRVRL PP | |
Greater than 90% as determined by SDS-PAGE. |
3897 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |