missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD20 / MS4A1 Protein
A DNA sequence encoding the Homo sapiens (Human) CD20 / MS4A1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9287-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_068769.2 | |
CD20 / MS4A1 Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
931 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
MS4A1 | |
Recombinant Protein | |
Yeast | |
IKISHFLKME SLNFIRAHTP YINIYNCEPA NPSEKNSPST QYCYSIQSLF LGILSVMLIF AFFQELVIAG IVENEWKRTC SRPKSNIVLL SAEEKKEQTI EIKEEVVGLT ETSSQPKNEE DIEIIPIQEE EEEETETNFP EPPQDQESSP IENDSSP |