missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD300LF / LMIR3 Protein

A DNA sequence encoding the Homo sapiens (Human) CD300LF / LMIR3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7829-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061041

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001276011.1
CD300LF / LMIR3 Protein
Research Use Only
CD300LF
Recombinant Protein
E. coli
TQITGPTTVN GLERGSLTVQ CVYRSGWETY LKWWCRGAIW RDCKILVKTS GSEQEVKRDR VSIKDNQKNR TFTVTMEDLM KTDADTYWCG IEKTGNDLGV TVQVTIDPAP VTQEETSSSP TLTGHHLDNR HKLLKLS
Greater than 90% as determined by SDS-PAGE.
146722
200 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers