missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD320 Protein

A DNA sequence encoding the Homo sapiens (Human) CD320, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8696-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16066001

  • 4074.00 SEK / 100µg


Estimated Shipment
11-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_001159367.1
CD320 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
51293
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CD320
Recombinant Protein
E. coli
SPLSTPTSAQ AAGPSSGSCP PTKFQCRTSG LCVPLTWRCD RDLDCSDGSD EEECRIEPCT QKGQCPPPPG LPCPCTGVSD CSGGTDKKLR NCSRLACLAG ELRCTLSDDC IPLTWRCDGH PDCPDSSDEL GCGTNEILPE GDATTMGPPV TLESVTSLRN ATTMGPPVTL ESVPSVGNAT SSSAGDQSGS PTAYGV
Videos
SDS
Documents