missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD3e / CD3 epsilon Protein
A DNA sequence encoding the Homo sapiens (Human) CD3e / CD3 epsilon, was expressed in the hosts and tags indicated.
1640.00 SEK - 18390.00 SEK
Specifications
Accession Number | NP_000724.1 |
---|---|
Gene ID (Entrez) | 916 |
Name | CD3e / CD3 epsilon Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035981
|
enQuireBio™
QP8691-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075981
|
enQuireBio™
QP8691-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025981
|
enQuireBio™
QP8691-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055981
|
enQuireBio™
QP8691-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065981
|
enQuireBio™
QP8691-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045981
|
enQuireBio™
QP8691-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000724.1 | |
CD3e / CD3 epsilon Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
916 | |
Research Use Only | |
CD3E | |
Recombinant Protein | |
E. coli | |
DGNEEMGGIT QTPYKVSISG TTVILTCPQY PGSEILWQHN DKNIGGDEDD KNIGSDEDHL SLKEFSELEQ SGYYVCYPRG SKPEDANFYL YLRARVCENC MEMDVMSVAT IVIVDICITG GLLLLVYYWS KNRKAKAKPV TRGAGAGGRQ RGQNKERPPP VPNPDYEPIR KGQRDLYSGL NQRRI | |
Greater than 90% as determined by SDS-PAGE. |