missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CEBPG / CEBP gamma Protein

A DNA sequence encoding the Homo sapiens (Human) CEBPG / CEBP gamma, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6946-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056820

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001239225.1
CEBPG / CEBP gamma Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
1054
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CEBPG
Recombinant Protein
E. coli
MSKISQQNST PGVNGISVIH TQAHASGLQQ VPQLVPAGPG GGGKAVAPSK QSKKSSPMDR NSDEYRQRRE RNNMAVKKSR LKSKQKAQDT LQRVNQLKEE NERLEAKIKL LTKELSVLKD LFLEHAHNLA DNVQSISTEN TTADGDNA
Videos
SDS
Documents