missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cell growth-regulating nucleolar Protein
A DNA sequence encoding the Homo sapiens (Human) Cell growth-regulating nucleolar protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9912-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001139197.1 | |
Cell growth-regulating nucleolar Protein | |
Research Use Only | |
LYAR | |
Recombinant Protein | |
Yeast | |
MVFFTCNACG ESVKKIQVEK HVSVCRNCEC LSCIDCGKDF WGDDYKNHVK CISEDQKYGG KGYEGKTHKG DIKQQAWIQK ISELIKRPNV SPKVRELLEQ ISAFDNVPRK KAKFQNWMKN SLKVHNESIL DQVWNIFSEA SNSEPVNKEQ DQRPLHPVAN PHAEISTKVP ASKVKDAVEQ QGEVKKNKRE RKEERQKKRK REKKELKLEN HQENSRNQKP KKRKKGQEAD LEAGGEEVPE ANGSAGKRSK KKKQRKDSAS EEEAHVGAGK RKRRHSEVET DSKKKKMKLP EHPEGGEPED DEAPAKGKFN WKGTIKAILK QAPDNEITIK KLRKKVLAQY YTVTDEHHRS EEELLVIFNK KISKNPTFKL LKDKVKLVK | |
Greater than 90% as determined by SDS-PAGE. |
55646 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |