missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Centromere protein 1
A DNA sequence encoding the Homo sapiens (Human) Centromere protein R, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7591-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001193668.1 | |
Centromere protein 1 | |
Research Use Only | |
ITGB3BP | |
Recombinant Protein | |
E. coli | |
MPVKRSLKLD GLLEENSFDP SKITRKKSVI TYSPTTGTCQ MSLFASPTSS EEQKHRNGLS NEKRKKLNHP SLTESKESTT KDNDEFMMLL SKVEKLSEEI MEIMQNLSSI QALEGSRELE NLIGISCASH FLKREMQKTK ELMTKVNKQK LFEKSTGLPH KASRHLDSYE FLKAILN | |
Greater than 80% as determined by SDS-PAGE. |
23421 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |