missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Centrosomal protein of 126 kDa Protein
A DNA sequence encoding the Homo sapiens (Human) Centrosomal protein of 126 kDa, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8097-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_065853.3 | |
Centrosomal protein of 126 kDa Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
57562 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
KIAA1377 | |
Recombinant Protein | |
E. coli | |
HKKMKYNIHE RNGVRFLKSI LKKESKYEHG YLKALIINQS FKFGNQKAAA IRDSIELTKE KGAEIPKTIK KLRWFDETSN IENNAENSHS LKNKTGTTQQ HSQQFHIQSG AG |