missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Centrosomal protein of 63 kDa Protein

A DNA sequence encoding the Homo sapiens (Human) Centrosomal protein of 63 kDa, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7898-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061471

  • 1580.00 SEK / 10µg


Estimated Shipment
02-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_001035842.1
Centrosomal protein of 63 kDa Protein
10 μg
Not determined.
Full Length of Isoform 3
Human
His-SUMO
Tris-based buffer, 50% glycerol
80254
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CEP63
Recombinant Protein
E. coli
MEALLEGIQN RGHGGGFLTS CEAELQELMK QIDIMVAHKK SEWEGRTHAL ETCLKIREQE LKSLRSQLDV THKEVGMLHQ QVEEHEKIKQ EMTMEYKQEL KKLHEELCIL KRSYEKLQKK QMREFRGNTK NHREDRSEIE RLTAKIEEFR QKSLDWEKQR LIYQQQVSSL EAQRKALAEQ SEIIQAQLVN RKQKLESVEL SSQSEIQHLS SKLERANDTI CANELEIERL TMRVNDLVGT SMTVLQEQQQ KEEKLRESEK LLEALQEEKR ELKAALQSQE NLIHEARIQK EKLQEKVKAT NTQHAVEAIS LESVSATCKQ LSQELMEKYE ELKRMEAHNN EYKAEIKKLK EQILQGEQSY SSALEGMKME ISHLTQELHQ RDITIASTKG SSSDMEKRLR AEMQKAEDKA VEHKEILDQL ESLKLENRHL SEMVMKLELG LHECSLPVSP LGSIATRFLE EEELRSHHIL ERLDAHIEEL KRESEKTVRQ FTALK
Videos
SDS
Documents