missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Claudin-6 Protein
A DNA sequence encoding the Homo sapiens(Human) Claudin-6, was expressed in the hosts and tags indicated.
4825.00 SEK - 19270.00 SEK
Specifications
Accession Number | NP_067018.2 |
---|---|
Gene ID (Entrez) | 9074 |
Name | Claudin-6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099111
|
enQuireBio™
QP9973-IV-100UG |
100 μg |
19270.00 SEK
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16019121
|
enQuireBio™
QP9973-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009121
|
enQuireBio™
QP9973-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_067018.2 | |
Claudin-6 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9074 | |
Research Use Only | |
CLDN6 | |
Recombinant Protein | |
E. coli | |
MASAGMQILG VVLTLLGWVN GLVSCALPMW KVTAFIGNSI VVAQVVWEGL WMSCVVQSTG QMQCKVYDSL LALPQDLQAA RA | |
Greater than 85% as determined by SDS-PAGE. |