missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Clavesin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Clavesin-2, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001010852.2 |
---|---|
Gene ID (Entrez) | 134829 |
Name | Clavesin-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16040171
|
enQuireBio™
QP7684-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060171
|
enQuireBio™
QP7684-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050171
|
enQuireBio™
QP7684-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070171
|
enQuireBio™
QP7684-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080171
|
enQuireBio™
QP7684-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090171
|
enQuireBio™
QP7684-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001010852.2 | |
Clavesin-2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
134829 | |
Research Use Only | |
CLVS2 | |
Recombinant Protein | |
E. coli | |
MTHLQAGLSP ETLEKARLEL NENPDTLHQD IQEVRDMVIT RPDIGFLRTD DAFILRFLRA RKFHHFEAFR LLAQYFEYRQ QNLDMFKSFK ATDPGIKQAL KDGFPGGLAN LDHYGRKILV LFAANWDQSR YTLVDILRAI LLSLEAMIED PELQVNGFVL IIDWSNFTFK QASKLTPSML RLAIEGLQDS FPARFGGIHF VNQPWYIHAL YTVIRPFLKE KTRKRIFLHG NNLNSLHQLI HPEILPSEFG GMLPPYDMGT WARTLLDHEY DDDSEYNVDS YSMPVKEVEK ELSPKSMKRS QSVVDPTVLK RMDKNEEENM QPLLSLD | |
Greater than 90% as determined by SDS-PAGE. |