missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CNGA4 Protein

A DNA sequence encoding the Homo sapiens (Human) CNGA4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9966-iv-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16059101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001032406.1
CNGA4 Protein
10 μg
Not determined.
Full Length Transmembrane Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
1262
Greater than 85% as determined by SDS-PAGE.
Research Use Only
CNGA4
Recombinant Protein
Invitro-E. coli
MSQDTKVKTT ESSPPAPSKA RKLLPVLDPS GDYYYWWLNT MVFPVMYNLI ILVCRACFPD LQHGYLVAWL VLDYTSDLLY LLDMVVRFHT GFLEQGILVV DKGRISSRYV RTWSFFLDLA SLMPTDVVYV RLGPHTPTLR LNRFLRAPRL FEAFDRTETR TAYPNAFRIA KLMLYIFVVI HWNSCLYFAL SRYLGFGRDA WVYPDPAQPG FERLRRQYLY SFYFSTLILT TVGDTPPPAR EEEYLFMVGD FLLAVMGFAT IMGSMSSVIY NMNTADAAFY PDHALVKKYM KLQHVNRKLE RRVIDWYQHL QINKKMTNEV AILQHLPERL RAEVAVSVHL STLSRVQIFQ NCEASLLEEL VLKLQPQTYS PGEYVCRKGD IGQEMYIIRE GQLAVVADDG ITQYAVLGAG LYFGEISIIN IKGNMSGNRR TANIKSLGYS DLFCLSKEDL REVLSEYPQA QTIMEEKGRE ILLKMNKLDV NAEAAEIALQ EATESRLRGL DQQLDDLQTK FARLLAELES SALKIAYRIE RLEWQTREWP MPEDLAEADD EGEPEEGTSK DEEGRASQEG PPGPE
Videos
SDS
Documents