missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNGA4 Protein
A DNA sequence encoding the Homo sapiens (Human) CNGA4, was expressed in the hosts and tags indicated.
6820.00 SEK - 21030.00 SEK
Specifications
Accession Number | NP_001032406.1 |
---|---|
Gene ID (Entrez) | 1262 |
Name | CNGA4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059101
|
enQuireBio™
QP9966-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069101
|
enQuireBio™
QP9966-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049101
|
enQuireBio™
QP9966-IV-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001032406.1 | |
CNGA4 Protein | |
Not determined. | |
Full Length Transmembrane Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1262 | |
Research Use Only | |
CNGA4 | |
Recombinant Protein | |
E. coli | |
MSQDTKVKTT ESSPPAPSKA RKLLPVLDPS GDYYYWWLNT MVFPVMYNLI ILVCRACFPD LQHGYLVAWL VLDYTSDLLY LLDMVVRFHT GFLEQGILVV DKGRISSRYV RTWSFFLDLA SLMPTDVVYV RLGPHTPTLR LNRFLRAPRL FEAFDRTETR TAYPNAFRIA KLMLYIFVVI HWNSCLYFAL SRYLGFGRDA WVYPDPAQPG FERLRRQYLY SFYFSTLILT TVGDTPPPAR EEEYLFMVGD FLLAVMGFAT IMGSMSSVIY NMNTADAAFY PDHALVKKYM KLQHVNRKLE RRVIDWYQHL QINKKMTNEV AILQHLPERL RAEVAVSVHL STLSRVQIFQ NCEASLLEEL VLKLQPQTYS PGEYVCRKGD IGQEMYIIRE GQLAVVADDG ITQYAVLGAG LYFGEISIIN IKGNMSGNRR TANIKSLGYS DLFCLSKEDL REVLSEYPQA QTIMEEKGRE ILLKMNKLDV NAEAAEIALQ EATESRLRGL DQQLDDLQTK FARLLAELES SALKIAYRIE RLEWQTREWP MPEDLAEADD EGEPEEGTSK DEEGRASQEG PPGPE | |
Greater than 85% as determined by SDS-PAGE. |