missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Coagulation Factor II / FII / F2 Protein
A DNA sequence encoding the Homo sapiens (Human) Coagulation Factor II / FII / F2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6009-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000497.1 | |
Coagulation Factor II / FII / F2 Protein | |
Research Use Only | |
F2 | |
Recombinant Protein | |
Yeast | |
ANTFLEEVRK GNLERECVEE TCSYEEAFEA LESSTATDVF WAKYTACETA RTPRDKLAAC LEGNCAEGLG TNYRGHVNIT RSGIECQLWR SRYPHKPEIN STTHPGADLQ ENFCRNPDSS TTGPWCYTTD PTVRRQECSI PVCGQDQVTV AMTPRSEGSS VNLSPPLEQC VPDRGQQYQG RLAVTTHGLP CLAWASAQAK ALSKHQDFNS AVQLVENFCR NPDGDEEGVW CYVAGKPGDF GYCDLNYCEE AVEEETGDGL DEDSDRAIEG RTATSEYQTF FNPRTFGSGE ADCGLRPLFE KKSLEDKTER ELLESYIDGR IVEGSDAEIG MSPWQVMLFR KSPQELLCGA SLISDRWVLT AAHCLLYPPW DKNFTENDLL VRIGKHSRTR YERNIEKISM LEKIYIHPRY NWRENLDRDI ALMKLKKPVA FSDYIHPVCL PDRETAASLL QAGYKGRVTG WGNLKETWTA NVGKGQPSVL QVVNLPIVER PVCKDSTRIR ITDNMFCAGY KPDEGKRGDA CEGDSGGPFV MKSPFNNRWY QMGIVSWGEG CDRDGKYGFY THVFRLKKWI QKVIDQFGE | |
Greater than 90% as determined by SDS-PAGE. |
2147 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |