Läs mer
enQuireBio™ Recombinant Human Collagen alpha-1(IV) chain Protein
Produktkod. 16046101
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16046101
Brand: enQuireBio™ QP8716ec1mg
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(IV) chain, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_001290039.1
Gen-ID (Entrez) 1282
Namn Collagen alpha-1(IV) chain Protein
Kvantitet 1 mg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol COL4A1
Protein Längd Partial (see sequence information for more details).
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag GST
Sekvens GCAGSGCGKC DCHGVKGQKG ERGLPGLQGV IGFPGMQGPE GPQGPPGQKG DTGEPGLPGT KGTRGPPGAS GYPGNPGLPG IPGQDGPPGP PGIPGCNGTK GERGPLGPPG LPGFAGNPGP PGLPGMKGDP GEILGHVP
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.