missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-6(VI) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-6(VI) chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7431-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001096078.1 | |
Collagen alpha-6(VI) chain Protein | |
Research Use Only | |
COL6A6 | |
Recombinant Protein | |
Yeast | |
VDTEEADIYL LIDGSGSTQA TDFHEMKTFL SEVVGMFNIA PHKVRVGAVQ YADSWDLEFE INKYSNKQDL GKAIENIRQM GGNTNTGAAL NFTLSLLQKA KKQRGNKVPC HLVVLTNGMS KDSILEPANR LREEHIRVYA IGIKEANQTQ LREIAGEEKR VYYVHDFDAL KDIRNQVVQE ICTEEACKEM KADIMFLVDS SGSIGPENFS KMKTFMKNLV SKSQIGPDRV QIGVVQFSDI NKEEFQLNRF MSQSDISNAI DQMAHIGQTT LTGSALSFVS QYFSPTKGAR PNIRKFLILI TDGEAQDIVK EPAVVLRQEG VIIYSVGVFG SNVTQLEEIS GRPEMVFYVE NFDILQRIED DLVFGICSPR EECKRIEVLD VVFVIDSSGS IDYDEYNIMK DFMIGLVKKA DVGKNQVRFG ALKYADDPEV LFYLDDFGTK LEVISVLQND QAMGGSTYTA EALGFSDHMF TEARGSRLNK GVPQVLIVIT DGESHDADKL NATAKALRDK GILVLAVGID GANPVELLAM AGSSDKYFFV ETFGGLKGIF SDV | |
Greater than 90% as determined by SDS-PAGE. |
131873 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |