missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagenase 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Collagenase 3, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_002418.1 |
---|---|
Gene ID (Entrez) | 4322 |
Name | Collagenase 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16002790
|
enQuireBio™
QP6367-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022790
|
enQuireBio™
QP6367-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16012790
|
enQuireBio™
QP6367-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032790
|
enQuireBio™
QP6367-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042790
|
enQuireBio™
QP6367-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052790
|
enQuireBio™
QP6367-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002418.1 | |
Collagenase 3 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
4322 | |
Research Use Only | |
MMP13 | |
Recombinant Protein | |
E. coli | |
YNVFPRTLKW SKMNLTYRIV NYTPDMTHSE VEKAFKKAFK VWSDVTPLNF TRLHDGIADI MISFGIKEHG DFYPFDGPSG LLAHAFPPGP NYGGDAHFDD DETWTSSSKG YNLFLVAAHE FGHSLGLDHS KDPGALMFPI YTYTGKSHFM LPDDDVQGIQ SLYGPGDEDP NPKHPKTPDK CDPSLSLDAI TSLRGETMIF KDRFFWRLHP QQVDAELFLT KSFWPELPNR IDAAYEHPSH DLIFIFRGRK FWALNGYDIL EGYPKKISEL GLPKEVKKIS AAVHFEDTGK TLLFSGNQVW RYDDTNHIMD KDYPRLIEED FPGIGDKVDA VYEKNGYIYF FNGPIQFEYS IWSNRIVRVM PANSILWC | |
Greater than 80% as determined by SDS-PAGE. |