missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Complement C4-B Protein
A DNA sequence encoding the Homo sapiens (Human) Complement C4-B, was expressed in the hosts and tags indicated.
2005.00 SEK - 21365.00 SEK
Specifications
Accession Number | NP_001002029.3 |
---|---|
Gene ID (Entrez) | 100293534 |
Name | Complement C4-B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036970
|
enQuireBio™
QP6998-YE-100UG |
100 μg |
7265.00 SEK
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076970
|
enQuireBio™
QP6998-YE-500UG |
500 μg |
19700.00 SEK
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046970
|
enQuireBio™
QP6998-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066970
|
enQuireBio™
QP6998-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086970
|
enQuireBio™
QP6998-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056970
|
enQuireBio™
QP6998-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001002029.3 | |
Complement C4-B Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
100293534 | |
Research Use Only | |
C4B | |
Recombinant Protein | |
Yeast | |
EAPKVVEEQE SRVHYTVCIW RNGKVGLSGM AIADVTLLSG FHALRADLEK LTSLSDRYVS HFETEGPHVL LYFDSVPTSR ECVGFEAVQE VPVGLVQPAS ATLYDYYNPE RRCSVFYGAP SKSRLLATLC SAEVCQCAEG KCPRQRRALE RGLQDEDGYR MKFACYYPRV EYGFQVKVLR EDSRAAFRLF ETKITQVLHF TKDVKAAANQ MRNFLVRASC RLRLEPGKEY LIMGLDGATY DLEGHPQYLL DSNSWIEEMP SERLCRSTRQ RAACAQLNDF LQEYGTQGCQ V | |
Greater than 90% as determined by SDS-PAGE. |