missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CPEB1 Protein

A DNA sequence encoding the Homo sapiens (Human) CPEB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8045-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062351

  • 26990.00 SEK / 1mg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001073001.1
CPEB1 Protein
Research Use Only
CPEB1
Recombinant Protein
E. coli
MLFPTSAQES SRGLPDANDL CLGLQSLSLT GWDRPWSTQD SDSSAQSSTH SVLSMLHNPL GNVLGKPPLS FLPLDPLGSD LVDKFPAPSV RGSRLDTRPI LDSRSSSPSD SDTSGFSSGS DHLSDLISSL RISPPLPFLS LSGGGPRDPL KMGVGSRMDQ EQAALAAVTP SPTSASKRWP GASVWPSWDL LEAPKDPFSI EREARLHRQA AAVNEATCTW SGQLPPRNYK NPIYSCKVFL GGVPWDITEA GLVNTFRVFG SLSVEWPGKD GKHPRCPPKG YVYLVFELEK SVRSLLQACS HDPLSPDGLS EYYFKMSSRR MRCKEVQVIP WVLADSNFVR SPSQRLDPSR TVFVGALHGM LNAEALAAIL NDLFGGVVYA GIDTDKHKYP IGSGRVTFNN QRSYLKAVSA AFVEIKTTKF TKKVQIDPYL EDSLCHICSS QPGPFFCRDQ VCFKYFCRSC WHWRHSMEGL RHHSPLMRNQ KNRDSS
Greater than 90% as determined by SDS-PAGE.
64506
1 mg
Not determined.
Full Length of isoform 4
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers