missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cripto / TDGF1 Protein
A DNA sequence encoding the Homo sapiens (Human) Cripto / TDGF1, was expressed in the hosts and tags indicated.
1640.00 SEK - 19050.00 SEK
Specifications
Accession Number | NP_001167607.1 |
---|---|
Gene ID (Entrez) | 6997 |
Name | Cripto / TDGF1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036041
|
enQuireBio™
QP8703-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076041
|
enQuireBio™
QP8703-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026041
|
enQuireBio™
QP8703-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056041
|
enQuireBio™
QP8703-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066041
|
enQuireBio™
QP8703-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046041
|
enQuireBio™
QP8703-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001167607.1 | |
Cripto / TDGF1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6997 | |
Research Use Only | |
TDGF1 | |
Recombinant Protein | |
E. coli | |
GHQEFARPSR GYLAFRDDSI WPQEEPAIRP RSSQRVPPMG IQHSKELNRT CCLNGGTCML GSFCACPPSF YGRNCEHDVR KENCGSVPHD TWLPKKCSLC KCWHGQLRCF PQAFLPGCD | |
Greater than 90% as determined by SDS-PAGE. |