missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cripto / TDGF1 Protein
A DNA sequence encoding the Homo sapiens (Human) Cripto / TDGF1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8703-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001167607.1 | |
Cripto / TDGF1 Protein | |
Research Use Only | |
TDGF1 | |
Recombinant Protein | |
E. coli | |
GHQEFARPSR GYLAFRDDSI WPQEEPAIRP RSSQRVPPMG IQHSKELNRT CCLNGGTCML GSFCACPPSF YGRNCEHDVR KENCGSVPHD TWLPKKCSLC KCWHGQLRCF PQAFLPGCD | |
Greater than 90% as determined by SDS-PAGE. |
6997 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |