missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclophilin A / PPIA / CYPA Protein
A DNA sequence encoding the Homo sapiens (Human) Cyclophilin A / PPIA / CYPA, was expressed in the hosts and tags indicated.
3490.00 SEK - 20095.00 SEK
Specifications
Accession Number | NP_001287910.1 |
---|---|
Gene ID (Entrez) | 5478 |
Name | Cyclophilin A / PPIA / CYPA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16065431
|
enQuireBio™
QP8588-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005441
|
enQuireBio™
QP8588-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055431
|
enQuireBio™
QP8588-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085431
|
enQuireBio™
QP8588-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095431
|
enQuireBio™
QP8588-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075431
|
enQuireBio™
QP8588-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001287910.1 | |
Cyclophilin A / PPIA / CYPA Protein | |
Not determined. | |
Full Length | |
Human | |
Untagged | |
Tris-based buffer, 50% glycerol |
5478 | |
Research Use Only | |
PPIA | |
Recombinant Protein | |
E. coli | |
VNPTVFFDIA VDGEPLGRVS FELFADKVPK TAENFRALST GEKGFGYKGS CFHRIIPGFM CQGGDFTRHN GTGGKSIYGE KFEDENFILK HTGPGILSMA NAGPNTNGSQ FFICTAKTEW LDGKHVVFGK VKEGMNIVEA MERFGSRNGK TSKKITIADC GQLE | |
Greater than 90% as determined by SDS-PAGE. |