missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cystatin B / CSTB Protein
A DNA sequence encoding the Homo sapiens (Human) Cystatin B / CSTB, was expressed in the hosts and tags indicated.
1580.00 SEK - 19050.00 SEK
Specifications
Accession Number | NP_000091.1 |
---|---|
Gene ID (Entrez) | 1476 |
Name | Cystatin B / CSTB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043951
|
enQuireBio™
QP8313-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083951
|
enQuireBio™
QP8313-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033951
|
enQuireBio™
QP8313-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063951
|
enQuireBio™
QP8313-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073951
|
enQuireBio™
QP8313-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053951
|
enQuireBio™
QP8313-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000091.1 | |
Cystatin B / CSTB Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1476 | |
Research Use Only | |
CSTB | |
Recombinant Protein | |
E. coli | |
MMCGAPSATQ PATAETQHIA DQVRSQLEEK ENKKFPVFKA VSFKSQVVAG TNYFIKVHVG DEDFVHLRVF QSLPHENKPL TLSNYQTNKA KHDELTYF | |
Greater than 90% as determined by SDS-PAGE. |