missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytochrome c-type heme lyase Protein
A DNA sequence encoding the Homo sapiens (Human) Cytochrome c-type heme lyase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6149-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001116080.1 | |
Cytochrome c-type heme lyase Protein | |
Research Use Only | |
HCCS | |
Recombinant Protein | |
E. coli | |
MGLSPSAPAV AVQASNASAS PPSGCPMHEG KMKGCPVNTE PSGPTCEKKT YSVPAHQERA YEYVECPIRG TAAENKENLD PSNLMPPPNQ TPAPDQPFAL STVREESSIP RADSEKKWVY PSEQMFWNAM LKKGWKWKDE DISQKDMYNI IRIHNQNNEQ AWKEILKWEA LHAAECPCGP SLIRFGGKAK EYSPRARIRS WMGYELPFDR HDWIINRCGT EVRYVIDYYD GGEVNKDYQF TILDVRPALD SLSAVWDRMK VAWWRWTS | |
Greater than 80% as determined by SDS-PAGE. |
3052 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |