missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytokeratin 10 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytokeratin 10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9239-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000412.3 | |
Cytokeratin 10 Protein | |
Research Use Only | |
KRT10 | |
Recombinant Protein | |
Yeast | |
EQLAEQNRKD AEAWFNEKSK ELTTEIDNNI EQISSYKSEI TELRRNVQAL EIELQSQLAL KQSLEASLAE TEGRYCVQLS QIQAQISALE EQLQQIRAET ECQNTEYQQL LDIKIRLE | |
Greater than 90% as determined by SDS-PAGE. |
3858 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |