missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytokeratin, Acidic Protein
A DNA sequence encoding the Homo sapiens (Human) Cytokeratin, Acidic, was expressed in the hosts and tags indicated.
2005.00 SEK - 21365.00 SEK
Specifications
Accession Number | NP_778253.2 |
---|---|
Gene ID (Entrez) | 374454 |
Name | Cytokeratin, Acidic Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16000741
|
enQuireBio™
QP7778-YE-100UG |
100 μg |
7265.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040741
|
enQuireBio™
QP7778-YE-500UG |
500 μg |
19700.00 SEK
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010741
|
enQuireBio™
QP7778-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030741
|
enQuireBio™
QP7778-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050741
|
enQuireBio™
QP7778-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020741
|
enQuireBio™
QP7778-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_778253.2 | |
Cytokeratin, Acidic Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
374454 | |
Research Use Only | |
KRT77 | |
Recombinant Protein | |
Yeast | |
MSHQFSSQSA FSSMSRRVYS TSSSAGSGGG SPAVGSVCYA RGRCGGGGYG IHGRGFGSRS LYNLGGSRSI SINLMGRSTS GFCQGGGVGG FGGGRGFGVG STGAGGFGGG GFGGAGFGTS NFGLGGFGPY CPPGGIQEVT INQSLLEPLH LEVDPEIQRI KTQEREQIMV LNNKFASFID KVRFLEQQNQ VLQTKWELLQ QVNTSTGTNN LEPLLENYIG DLRRQVDLLS AEQMRQNAEV RSMQDVVEDY KSKYEDEINK RTGSENDFVV LKKDVDAAYV SKVDLESRVD TLTGEVNFLK YLFLTELSQV QTHISDTNVI LSMDNNRSLD LDSIIDAVRT QYELIAQRSK DEAEALYQTK YQELQITAGR HGDDLKNSKM EIAELNRTVQ RLQAEISNVK KQIEQMQSLI SDAEERGEQA LQDAWQKLQD LEEALQQSKE ELARLLRDYQ AMLGVKLSLD VEIATYRQLL EGEESRMSGE LQSHVSISVQ NSQVSVNGGA GGGGSYGSGG YGGGSGGGYG GGRSYRGGGA RGRSGGGYGS GCGGGGGSYG GSGRSGRGSS RVQIIQTSTN TSHRRILE | |
Greater than 90% as determined by SDS-PAGE. |