missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DBI / ACBD1 Protein
A DNA sequence encoding the Homo sapiens (Human) DBI / ACBD1, was expressed in the hosts and tags indicated.
2405.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001073331.1 |
---|---|
Gene ID (Entrez) | 1622 |
Name | DBI / ACBD1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933308
|
enQuireBio™
QP1027-EC-10UG |
10 μg |
2405.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923308
|
enQuireBio™
QP1027-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943308
|
enQuireBio™
QP1027-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953308
|
enQuireBio™
QP1027-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15963308
|
enQuireBio™
QP1027-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15973308
|
enQuireBio™
QP1027-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001073331.1 | |
DBI / ACBD1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1622 | |
Research Use Only | |
DBI | |
Recombinant Protein | |
E. coli | |
SQAEFEKAAE EVRHLKTKPS DEEMLFIYGH YKQATVGDIN TERPGMLDFT GKAKWDAWNE LKGTSKEDAM KAYINKVEEL KKKYGI | |
Greater than 90% as determined by SDS-PAGE. |