missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Recombinant Human Desmoglein-4 Fc Chimera Protein

Produktkod. 18768649
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18768649 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18768649 Leverantör R&D Systems™ Leverantörsnummer 9230DM100
 Begränsad artikel
Beräknad leverans: 27-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Desmoglein-4 Fc Chimera Protein is derived from HEK293. The Recombinant Human Desmoglein-4 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Beställningsinfo

The product is shipped at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Tillträdesnummer Q86SJ6
För användning med (applikation) Bioactivity
Formulering Lyophilized from a 0.2μm filtered solution in PBS and NaCl
Gen-ID (Entrez) 147409
Molekylvikt (g/mol) 95 to 115 kDa, reducing conditions
Namn Cad13, Cadherin family member 13, CDGF13, CDHF13, CDHF13CDH family member 13, desmoglein 4, Desmoglein4, DSG4, LAHdesmoglein-4
Kvantitet 100 μg
Källa Human embryonic kidney cell,HEK293-derived human Desmoglein-4 protein, (N-terminus) Human Desmoglein-4 (Glu24-Ala631) Accession &Num; Q86SJ6 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Förvaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatorisk status RUO
Endotoxinkoncentration <0.10 EU per 1μg of protein by LAL method
Biologisk aktivitet 0.4 to 2.4μg/mL (ED 50)
Produkttyp Recombinant protein
Konjugera Unconjugated
Art Human
Rekombinant Recombinant
Protein Tag Yes
Produktlinje R&D Systems
Renhet eller kvalitetsklass >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.