missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Diamine acetyltransferase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Diamine acetyltransferase 1, was expressed in the hosts and tags indicated.
1580.00 SEK - 18390.00 SEK
Specifications
Accession Number | NP_002961.1 |
---|---|
Gene ID (Entrez) | 6303 |
Name | Diamine acetyltransferase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16054541
|
enQuireBio™
QP8424-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094541
|
enQuireBio™
QP8424-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044541
|
enQuireBio™
QP8424-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074541
|
enQuireBio™
QP8424-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084541
|
enQuireBio™
QP8424-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064541
|
enQuireBio™
QP8424-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002961.1 | |
Diamine acetyltransferase 1 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6303 | |
Research Use Only | |
SAT1 | |
Recombinant Protein | |
E. coli | |
VIRPATAADC SDILRLIKEL AKYEYMEEQV ILTEKDLLED GFGEHPFYHC LVAEVPKEHW TPEGHSIVGF AMYYFTYDPW IGKLLYLEDF FVMSDYRGFG IGSEILKNLS QVAMRCRCSS MHFLVAEWNE PSINFYKRRG ASDLSSEEGW RLFKIDKEYL LKMATEE | |
Greater than 90% as determined by SDS-PAGE. |