missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dihydropyrimidinase-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Dihydropyrimidinase-related protein 1, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001014809.1 |
---|---|
Gene ID (Entrez) | 1400 |
Name | Dihydropyrimidinase-related protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039760
|
enQuireBio™
QP7611-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059760
|
enQuireBio™
QP7611-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049760
|
enQuireBio™
QP7611-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069760
|
enQuireBio™
QP7611-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079760
|
enQuireBio™
QP7611-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089760
|
enQuireBio™
QP7611-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001014809.1 | |
Dihydropyrimidinase-related protein 1 | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1400 | |
Research Use Only | |
CRMP1 | |
Recombinant Protein | |
E. coli | |
MSYQGKKSIP HITSDRLLIK GGRIINDDQS LYADVYLEDG LIKQIGENLI VPGGVKTIEA NGRMVIPGGI DVNTYLQKPS QGMTAADDFF QGTRAALVGG TTMIIDHVVP EPGSSLLTSF EKWHEAADTK SCCDYSLHVD ITSWYDGVRE ELEVLVQDKG VNSFQVYMAY KDVYQMSDSQ LYEAFTFLKG LGAVILVHAE NGDLIAQEQK RILEMGITGP EGHALSRPEE LEAEAVFRAI TIAGRINCPV YITKVMSKSA ADIIALARKK GPLVFGEPIA ASLGTDGTHY WSKNWAKAAA FVTSPPLSPD PTTPDYLTSL LACGDLQVTG SGHCPYSTAQ KAVGKDNFTL IPEGVNGIEE RMTVVWDKAV ATGKMDENQF VAVTSTNAAK IFNLYPRKGR IAVGSDADVV IWDPDKLKTI TAKSHKSAVE YNIFEGMECH GSPLVVISQG KIVFEDGNIN VNKGMGRFIP RKAFPEHLYQ RVKIRNKVFG LQGVSRGMYD GPVYEVPATP KYATPAPSAK SSPSKHQPPP IRNLHQSNFS LSGAQIDDNN PRRTGHRIVA PPGGRSNITS LG | |
Greater than 90% as determined by SDS-PAGE. |