missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dihydropyrimidinase-related protein 1

A DNA sequence encoding the Homo sapiens (Human) Dihydropyrimidinase-related protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7611-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16059760

  • 18585.00 SEK / 1mg


Estimated Shipment
10-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_001014809.1
Dihydropyrimidinase-related protein 1
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
1400
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CRMP1
Recombinant Protein
E. coli
MSYQGKKSIP HITSDRLLIK GGRIINDDQS LYADVYLEDG LIKQIGENLI VPGGVKTIEA NGRMVIPGGI DVNTYLQKPS QGMTAADDFF QGTRAALVGG TTMIIDHVVP EPGSSLLTSF EKWHEAADTK SCCDYSLHVD ITSWYDGVRE ELEVLVQDKG VNSFQVYMAY KDVYQMSDSQ LYEAFTFLKG LGAVILVHAE NGDLIAQEQK RILEMGITGP EGHALSRPEE LEAEAVFRAI TIAGRINCPV YITKVMSKSA ADIIALARKK GPLVFGEPIA ASLGTDGTHY WSKNWAKAAA FVTSPPLSPD PTTPDYLTSL LACGDLQVTG SGHCPYSTAQ KAVGKDNFTL IPEGVNGIEE RMTVVWDKAV ATGKMDENQF VAVTSTNAAK IFNLYPRKGR IAVGSDADVV IWDPDKLKTI TAKSHKSAVE YNIFEGMECH GSPLVVISQG KIVFEDGNIN VNKGMGRFIP RKAFPEHLYQ RVKIRNKVFG LQGVSRGMYD GPVYEVPATP KYATPAPSAK SSPSKHQPPP IRNLHQSNFS LSGAQIDDNN PRRTGHRIVA PPGGRSNITS LG
Videos
SDS
Documents