missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dim2 / TXNL4B Protein
A DNA sequence encoding the Homo sapiens (Human) Dim2 / TXNL4B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8094-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001135789.1 | |
Dim2 / TXNL4B Protein | |
Research Use Only | |
TXNL4B | |
Recombinant Protein | |
E. coli | |
MSFLLPKLTS KKEVDQAIKS TAEKVLVLRF GRDEDPVCLQ LDDILSKTSS DLSKMAAIYL VDVDQTAVYT QYFDISYIPS TVFFFNGQHM KVDYGSPDHT KFVGSFKTKQ DFIDLIEVIY RGAMRGKLIV QSPIDPKNIP KYDLLYQDI | |
Greater than 80% as determined by SDS-PAGE. |
54957 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |