missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Recombinant Human Dkk-1 Biotinylated Protein

Produktkod. 18749317
Change view
Klicka för att se tillgängliga alternativ
Formulering:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Formulering
18749317 Lyophilized from a 0.2μm filtered solution in PBS
18716109 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18749317 Leverantör R&D Systems™ Leverantörsnummer BT5439010/CF
 Begränsad artikel
Beräknad leverans: 03-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Dkk-1 Biotinylated Protein is derived from Sf 21 (baculovirus). The Recombinant Human Dkk-1 Biotinylated Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer O94907
Formulering Lyophilized from a 0.2μm filtered solution in PBS
Gen-ID (Entrez) 22943
Molekylvikt (g/mol) 26kDa (unlabeled)
Namn Dkk-1
Beredningsmetod Reconstitute at 100μg/mL in PBS.
Kvalitetskontrolltestning SDS-PAGE: 33-38kDa, reducing conditions
Kvantitet 10 μg
Källa Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Dkk-1 protein Thr32-His266
Förvaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatorisk status RUO
Uppsägning Thr32
Utan tillsatser Carrier Protein
Strukturell form Monomer, biotinylated via sugars
Konjugera Biotin
Rekombinant Recombinant
Produktlinje R&D Systems
Renhet eller kvalitetsklass 95%, by SDS-PAGE with silver staining
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.