missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA replication licensing factor MCM2 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA replication licensing factor MCM2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6349-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004517.2 | |
DNA replication licensing factor MCM2 Protein | |
Research Use Only | |
MCM2 | |
Recombinant Protein | |
E. coli | |
AESSESFTMA SSPAQRRRGN DPLTSSPGRS SRRTDALTSS PGRDLPPFED ESEGLLGTEG PLEEEEDGEE LIGDGMERDY RAIPELDAYE AEGLALDDED VEELTASQRE AAERAMRQRD REAGRGLGRM RRGLLYDSDE EDEERPARKR RQVERATEDG EEDEEMIESI ENLEDLKGHS VREWVSMAGP RLEIHHRFKN FLRTHVDSHG HNVFKERISD MCKENRESLV VNYEDLAARE HVLAYFLPEA PAELLQIFDE AALEVVLAMY PKYDRITNHI HVRISHLPLV EELRSLRQLH LNQLIRTSGV VTSCTGVLPQ LSMVKYNCNK CNFVLGPFCQ SQNQEVKPGS CPECQSAGPF EVNMEETIYQ NYQRIRIQES PGKVAAGRLP RSKDAILLAD LVDSCKPGDE IELTGIYHNN YDGSLNTANG FPVFATVILA NHVAKKDNKV AVGELTDEDV KMITSLSKDQ QIGEKIFASI APSIYGHEDI KRGLALALFG GEPKNPGGKH KVRGDINVLL CGDPGTAKSQ FLKYIEKVSS RAIFTTGQGA SAVGLTAYVQ RHPVSREWTL EAGALVLADR GVCLIDEFDK MNDQDRTSIH EAMEQQSISI SKAGIVTSLQ ARCTVIAAAN PIGGRYDPSL TFSENVDLTE PIISRFDILC VVRDTVDPVQ DEMLARFVVG SHVRHHPSNK EEEGLANGSA AEPAMPNTYG VEPLPQEVLK KYIIYAKERV HPKLNQMDQD KVAKMYSDLR KESMATGSIP ITVRHIESMI RMAEAHARIH LRDYVIEDDV NMAIRVMLES FIDTQKFSVM RSMRKTFARY LSFRRDNNEL LLFILKQLVA EQVTYQRNRF GAQQDTIEVP EKDLVDKARQ INIHNLSAFY DSELFRMNKF SHDLKRKMIL QQF | |
Greater than 90% as determined by SDS-PAGE. |
4171 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |