missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DnaJ homolog subfamily B member 1 Protein
A DNA sequence encoding the Homo sapiens (Human) DnaJ homolog subfamily B member 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8461-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001287843.1 | |
DnaJ homolog subfamily B member 1 Protein | |
Research Use Only | |
DNAJB1 | |
Recombinant Protein | |
E. coli | |
MGKDYYQTLG LARGASDEEI KRAYRRQALR YHPDKNKEPG AEEKFKEIAE AYDVLSDPRK REIFDRYGEE GLKGSGPSGG SGGGANGTSF SYTFHGDPHA MFAEFFGGRN PFDTFFGQRN GEEGMDIDDP FSGFPMGMGG FTNVNFGRSR SAQEPARKKQ DPPVTHDLRV SLEEIYSGCT KKMKISHKRL NPDGKSIRNE DKILTIEVKK GWKEGTKITF PKEGDQTSNN IPADIVFVLK DKPHNIFKRD GSDVIYPARI SLREALCGCT VNVPTLDGRT IPVVFKDVIR PGMRRKVPGE GLPLPKTPEK RGDLIIEFEV IFPERIPQTS RTVLEQVLPI | |
Greater than 90% as determined by SDS-PAGE. |
3337 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |